α,2-Dibromo-5-fluorotoluene 98+%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Bromo-2-(bromomethyl)-4-fluorobenzene, 2-Bromo-5-fluorobenzyl bromide
PC4075-100G PC4075-25G PC4075-5G
APOSPC4075-100GEA 166 EUR
APOSPC4075-100G APOSPC4075-25G APOSPC4075-5G
α,2-Dibromo-5-fluorotoluene 98+%
α,2-Dibromo-5-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Boiling Pt: 89…90 °C/1mmHg
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3261
ADR: 8,II