2-Amino-5-fluoro-4-methylpyridine 98%
Supplier: Apollo Scientific
Warning
PC408287-5G PC408287-1G
APOSPC408287-5GEA 241 EUR
APOSPC408287-5G APOSPC408287-1G
2-Amino-5-fluoro-4-methylpyridine 98%
2-Amino-5-fluoro-4-methylpyridine
Formula: C₆H₇FN₂
MW: 126.13 g/mol
Melting Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD06659479
CAS Number: 301222-66-2