4,4-Difluoro-L-prolinamide hydrochloride
Supplier: Apollo Scientific
Synonyms: (S)-4,4-Difluoropyrrolidine-2-carboxamide hydrochloride
PC409000-1G PC409000-250MG
APOSPC409000-1GEA 190 EUR
APOSPC409000-1G APOSPC409000-250MG
4,4-Difluoro-L-prolinamide hydrochloride
4,4-Difluoro-L-prolinamide hydrochloride
Formula: C₅H₈F₂N₂O·HCl
MW: 186.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797014
CAS Number: 426844-51-1