N-(Piperidin-4-ylmethyl)-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride hydrate 95+%
Supplier: Apollo Scientific
PC409507-1G PC409507-5G
APOSPC409507-1GEA 1210 EUR
APOSPC409507-1G APOSPC409507-5G
N-(Piperidin-4-ylmethyl)-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride hydrate 95+%
N-(Piperidin-4-ylmethyl)-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride hydrate
Formula: C₁₅H₂₀ClF₃N₄O
MW: 364.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08567944
CAS Number: 1356114-70-9