1-{1-[2-(Trifluoromethyl)-1,6-naphthyridin-5-yl]piperidin-4-yl}methanamine 95+%
Supplier: Apollo Scientific
PC409508-1G PC409508-5G
APOSPC409508-1GEA 1210 EUR
APOSPC409508-1G APOSPC409508-5G
1-{1-[2-(Trifluoromethyl)-1,6-naphthyridin-5-yl]piperidin-4-yl}methanamine 95+%
1-{1-[2-(Trifluoromethyl)-1,6-naphthyridin-5-yl]piperidin-4-yl}methanamine
Formula: C₁₅H₁₇F₃N₄
Storage Temperature: Ambient
MDL Number: MFCD08566157
CAS Number: 909644-98-0