5-Piperazin-1-yl-2-(trifluoromethyl)-1,6-naphthyridine 95+%
Supplier: Apollo Scientific
PC409512-1G PC409512-10G PC409512-5G
APOSPC409512-1GEA 348 EUR
APOSPC409512-1G APOSPC409512-10G APOSPC409512-5G
5-Piperazin-1-yl-2-(trifluoromethyl)-1,6-naphthyridine 95+%
5-Piperazin-1-yl-2-(trifluoromethyl)-1,6-naphthyridine
Formula: C₁₃H₁₃F₃N₄
Storage Temperature: Ambient
MDL Number: MFCD08565762
CAS Number: 890302-17-7