1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea 98%
Supplier: Apollo Scientific
PC4097-10G PC4097-1G PC4097-5G
APOSPC4097-10GEA 495 EUR
APOSPC4097-10G APOSPC4097-1G APOSPC4097-5G
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea 98%
1-[4-Chloro-3-(trifluoromethyl)phenyl]-2-thiourea
Formula: C₈H₆ClF₃N₂S
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00041182
CAS Number: 207919-03-7