Diethyl tetrafluorosuccinate 97%
Supplier: Apollo Scientific
Warning
PC4102-5G PC4102-25G PC4102-1G
APOSPC4102-5GEA 111 EUR
APOSPC4102-5G APOSPC4102-25G APOSPC4102-1G
Diethyl tetrafluorosuccinate 97%
Diethyl tetrafluorosuccinate
Formula: C₈H₁₀F₄O₄
Boiling Pt: 78 °C/5mmHg
Density: 1.273 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015155
CAS Number: 377-71-9
EINECS: 206-821-2