5-Bromo-2-fluorobenzoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
PC4107-5G
PC4107-100G
PC4107-25G
APOSPC4107-5GEA
21.9
EUR
APOSPC4107-5G
APOSPC4107-100G
APOSPC4107-25G
5-Bromo-2-fluorobenzoic acid 97%
5-Bromo-2-fluorobenzoic acid
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol Melting Pt: 141…151 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143423 CAS Number: 146328-85-0 ADR: 6.1,III |