1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Difluoromethyl)-1H-pyrazole-3-carboxylic acid
PC411317-1G PC411317-25G PC411317-5G
APOSPC411317-1GEA 121 EUR
APOSPC411317-1G APOSPC411317-25G APOSPC411317-5G
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
Formula: C₅H₄F₂N₂O₂
MW: 162.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03420211
CAS Number: 925179-02-8