(±)-3,4-Difluoromandelic acid 97%
Supplier: Apollo Scientific
Warning
Synonyms: (±)-3,4-Difluorophenylglycolic acid
PC4116-1G PC4116-250MG PC4116-5G
APOSPC4116-1GEA 23.2 EUR
APOSPC4116-1G APOSPC4116-250MG APOSPC4116-5G
(±)-3,4-Difluoromandelic acid 97%
(±)-3,4-Difluoromandelic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00061299
CAS Number: 132741-29-8