(4-Fluorophenyl)acetone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorophenylacetone
PC4141L-100G PC4141L-25G PC4141L-5G
APOSPC4141L-100GEA 132 EUR
APOSPC4141L-100G APOSPC4141L-25G APOSPC4141L-5G
(4-Fluorophenyl)acetone 98%
(4-Fluorophenyl)acetone
Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 106…107 °C/18mmHg
Density: 1.139 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000362
CAS Number: 459-03-0
EINECS: 207-284-7