(3-Fluorophenyl)acetonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(3-Fluorophenyl)acetonitrile, 3-Fluorobenzyl cyanide
PC4143-100G
PC4143-25G
PC4143-5G
PC4143-10G
APOSPC4143-100GEA
210
EUR
APOSPC4143-100G
APOSPC4143-25G
APOSPC4143-5G
APOSPC4143-10G
(3-Fluorophenyl)acetonitrile 98%
(3-Fluorophenyl)acetonitrile
Formula:
C₈H₆FN Boiling Pt: 124…125 °C/10mmHg Density: 1.163 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001907 CAS Number: 501-00-8 EINECS: 207-918-2 UN: 3276 ADR: 6.1,III |