(4-Fluorophenyl)acetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile
PC4144-100G PC4144-25G
APOSPC4144-100GEA 29 EUR
APOSPC4144-100G APOSPC4144-25G
(4-Fluorophenyl)acetonitrile 98%
(4-Fluorophenyl)acetonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 119…120 °C/18mmHg
Density: 1.126 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001917
CAS Number: 459-22-3
EINECS: 207-286-8
UN: 3276
ADR: 6.1,III