4-Fluoro-DL-phenylalanine 98%
Supplier: Apollo Scientific
Warning
PC4149-1G PC4149-5G
APOSPC4149-1GEA 51 EUR
APOSPC4149-1G APOSPC4149-5G
4-Fluoro-DL-phenylalanine 98%
4-Fluoro-DL-phenylalanine
Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00002600
CAS Number: 51-65-0