2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98+%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-6-nitrobenzyl bromide, 1-(Bromomethyl)-2-fluoro-6-nitrobenzene
PC4155-5G PC4155-500MG PC4155-25G
APOSPC4155-5GEA 198 EUR
APOSPC4155-5G APOSPC4155-500MG APOSPC4155-25G
2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98+%
2-(Bromomethyl)-1-fluoro-3-nitrobenzene
Formula: C₇H₅BrFNO₂
MW: 234.02 g/mol
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00042918
CAS Number: 1958-93-6
EINECS: 217-801-8
UN: 3261
ADR: 8 II