1-(3-Fluorophenyl)-2-thiourea 98%
Supplier: Apollo Scientific
|
Danger
|
PC4175N-25G
PC4175N-5G
APOSPC4175N-25GEA
87
EUR
APOSPC4175N-25G
APOSPC4175N-5G
1-(3-Fluorophenyl)-2-thiourea 98%
1-(3-Fluorophenyl)-2-thiourea
Formula:
(C₆H₄F)NC(S)NH₂ Melting Pt: 114…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041167 CAS Number: 458-05-9 UN: 2811 ADR: 6.1,III |