Ethyl-4,4,4-trifluorobut-2-ynoate 96%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Ethyl 4,4,4-trifluorobut-2-ynoate
PC4177-5G
PC4177-1G
PC4177-10G
PC4177-25G
APOSPC4177-5GEA
200
EUR
APOSPC4177-5G
APOSPC4177-1G
APOSPC4177-10G
APOSPC4177-25G
Ethyl-4,4,4-trifluorobut-2-ynoate 96%
Ethyl-4,4,4-trifluorobut-2-ynoate
Formula:
C₆H₅F₃O₂ MW: 166.099 g/mol Boiling Pt: 96…98 °C Density: 1.162 g/cm³ (25 °C) Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192167 CAS Number: 79424-03-6 UN: 3272 ADR: 3,II |