Methyl-3,5-difluorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3,5-difluorobenzoate
PC4186-1G PC4186-25G PC4186-5G PC4186-100G
APOSPC4186-1GEA 14.7 EUR
APOSPC4186-1G APOSPC4186-25G APOSPC4186-5G APOSPC4186-100G
Methyl-3,5-difluorobenzoate 98%
Methyl 3,5-difluorobenzoate
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318160
CAS Number: 216393-55-4