2'-Fluoropropiophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: o-Fluoropropiophenone
PC4188-25G PC4188-5G
APOSPC4188-25GEA 46.4 EUR
APOSPC4188-25G APOSPC4188-5G
2'-Fluoropropiophenone 98%
2'-Fluoropropiophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 89…91 °C/12mmHg
Density: 1.102 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00009893
CAS Number: 446-22-0