2-Fluoropyridine 99%
Supplier: Apollo Scientific
Warning
PC4200-100G PC4200-250G PC4200-50G
APOSPC4200-100GEA 29 EUR
APOSPC4200-100G APOSPC4200-250G APOSPC4200-50G
2-Fluoropyridine 99%
2-Fluoropyridine
Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 124…126 °C
Density: 1.128 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006224
CAS Number: 372-48-5
EINECS: 206-757-5
UN: 1993
ADR: 3,III