Methyl-3,3-difluorocyclobutanecarboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3,3-difluorocyclobutanecarboxylate, Methyl 3,3-difluorocyclobutane-1-carboxylate
PC420000-25G
APOSPC420000-25GEA 215 EUR
APOSPC420000-25G
Methyl-3,3-difluorocyclobutanecarboxylate 98%
Methyl-3,3-difluorocyclobutanecarboxylate
Formula: C₆H₈F₂O₂
MW: 150.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015922
CAS Number: 1234616-13-7