2-Chloro-3-(difluoromethoxy)pyridine
Supplier: Apollo Scientific
Warning
PC421029-1G PC421029-250MG PC421029-5G
APOSPC421029-1GEA 48 EUR
APOSPC421029-1G APOSPC421029-250MG APOSPC421029-5G
2-Chloro-3-(difluoromethoxy)pyridine
2-Chloro-3-(difluoromethoxy)pyridine
Formula: C₆H₄ClF₂NO
MW: 179.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185847
CAS Number: 1206977-80-1