4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
PC421213-25G PC421213-1G PC421213-5G
APOSPC421213-25GEA 253 EUR
APOSPC421213-25G APOSPC421213-1G APOSPC421213-5G
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
MW: 258.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973757
CAS Number: 104460-70-0