2-Fluorostyrene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-2-vinylbenzene
PC4230-25G PC4230-5G PC4230-1G
APOSPC4230-25GEA 364 EUR
APOSPC4230-25G APOSPC4230-5G APOSPC4230-1G
2-Fluorostyrene 99%
2-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 46 °C/32mmHg
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013550
CAS Number: 394-46-7
EINECS: 206-896-1
UN: 1993
ADR: 3 III