5-Amino-3-(4-fluorophenyl)isoxazole 98%
Supplier: Apollo Scientific
Warning
PC4231-1G PC4231-250MG PC4231-5G
APOSPC4231-1GEA 219 EUR
APOSPC4231-1G APOSPC4231-250MG APOSPC4231-5G
5-Amino-3-(4-fluorophenyl)isoxazole 98%
5-Amino-3-(4-fluorophenyl)isoxazole
Formula: C₉H₇FN₂O
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD03407398
CAS Number: 81465-82-9