3-Fluorostyrene 98%
Supplier: Apollo Scientific
Warning
PC4240-25G PC4240-5G PC4240-1G PC4240-10G
APOSPC4240-25GEA 126 EUR
APOSPC4240-25G APOSPC4240-5G APOSPC4240-1G APOSPC4240-10G
3-Fluorostyrene 98%
3-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 31 °C/4mmHg
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000341
CAS Number: 350-51-6
EINECS: 206-504-9
UN: 1993
ADR: 3 III