3,5-Difluoro-4-methoxyaniline
Supplier: Apollo Scientific
Danger
PC4243-1G PC4243-5G PC4243-25G
APOSPC4243-1GEA 26.1 EUR
APOSPC4243-1G APOSPC4243-5G APOSPC4243-25G
3,5-Difluoro-4-methoxyaniline
3,5-Difluoro-4-methoxyaniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD04115910
CAS Number: 363-47-3
UN: 2811
ADR: 6.1 III