1-Bromo-2,6-difluoro-4-nitrobenzene 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1,3-difluoro-5-nitrobenzene
PC4245-1G PC4245-25G PC4245-5G
APOSPC4245-1GEA 48 EUR
APOSPC4245-1G APOSPC4245-25G APOSPC4245-5G
1-Bromo-2,6-difluoro-4-nitrobenzene 97+%
1-Bromo-2,6-difluoro-4-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.9864 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD04974121
CAS Number: 886762-62-5