4-Fluorostyrene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-4-vinylbenzene
PC4250-25G PC4250-5G
APOSPC4250-25GEA 97.3 EUR
APOSPC4250-25G APOSPC4250-5G
4-Fluorostyrene 95%
4-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 145…148 °C
Melting Pt: –34.5 °C
Density: 1.024 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000361
CAS Number: 405-99-2
EINECS: 206-975-0
UN: 1993
ADR: 3 III