4-Fluorothioanisole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorophenyl methyl sulphide, 1-Fluoro-4-(methylthio)benzene
PC4286-25G PC4286-5G
APOSPC4286-25GEA 29 EUR
APOSPC4286-25G APOSPC4286-5G
4-Fluorothioanisole 97%
4-Fluorothioanisole
Formula: C₇H₇FS
MW: 142.2 g/mol
Boiling Pt: 90 °C/31mmHg
Density: 1.167 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00040829
CAS Number: 371-15-3
EINECS: 206-733-4