6-(Trifluoromethyl)-1,3-dihydro-2H-benzimidazol-2-one
Supplier: Apollo Scientific
Synonyms: 5-(Trifluoromethyl)-1H-benzo[d]imidazol-2(3H)-one, 6-(Trifluoromethyl)-1H-benzo[d]imidazol-2-ol
PC430583-1G PC430583-250MG PC430583-2.5G PC430583-5G
APOSPC430583-1GEA 439 EUR
APOSPC430583-1G APOSPC430583-250MG APOSPC430583-2.5G APOSPC430583-5G
6-(Trifluoromethyl)-1,3-dihydro-2H-benzimidazol-2-one
6-(Trifluoromethyl)-1,3-dihydro-2H-benzimidazol-2-one
Formula: C₈H₅F₃N₂O
MW: 202.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663529
CAS Number: 133687-93-1