1-Bromo-2,3,5-trifluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,5-Trifluorobromobenzene
PC4309-5G PC4309-100G PC4309-25G
APOSPC4309-5GEA 21.9 EUR
APOSPC4309-5G APOSPC4309-100G APOSPC4309-25G
1-Bromo-2,3,5-trifluorobenzene 98%
1-Bromo-2,3,5-trifluorobenzene
Formula: C₆H₂BrF₃
Boiling Pt: 143 °C
Density: 1.777 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00012232
CAS Number: 133739-70-5
UN: 1993
ADR: 3,III