N-Methyl-2,2,2-trifluoroacetamide 95%
Supplier: Apollo Scientific
Warning
Synonyms: N-Methyltrifluoroacetamide, 2,2,2-Trifluoro-N-methylacetamide
PC4311-25G PC4311-5G PC4311-100G
APOSPC4311-25GEA 29 EUR
APOSPC4311-25G APOSPC4311-5G APOSPC4311-100G
N-Methyl-2,2,2-trifluoroacetamide 95%
N-Methyl-2,2,2-trifluoroacetamide
Formula: C₃H₄F₃NO
MW: 127.07 g/mol
Boiling Pt: 156…157 °C
Melting Pt: 49…51 °C
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00009670
CAS Number: 815-06-5