3-Fluorotoluene 99%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Fluoro-3-methylbenzene
PC4320-100G PC4320-1KG PC4320-500G
APOSPC4320-100GEA 21.9 EUR
APOSPC4320-100G APOSPC4320-1KG APOSPC4320-500G
3-Fluorotoluene 99%
3-Fluorotoluene
Intermediate for the production of Ciprofloxacin.
Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 114…116 °C
Melting Pt: –87 °C
Density: 0.991 g/cm³ (25 °C)
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00000339
CAS Number: 352-70-5
EINECS: 206-524-8
UN: 2388
ADR: 3,II