2-Chloro-5-(trifluoromethyl)benzylamine 95%
Supplier: Apollo Scientific
Danger
PC4327-25G PC4327-5G PC4327-1G
APOSPC4327-25GEA 157 EUR
APOSPC4327-25G APOSPC4327-5G APOSPC4327-1G
2-Chloro-5-(trifluoromethyl)benzylamine 95%
2-Chloro-5-(trifluoromethyl)benzylamine
Formula: C₈H₇ClF₃N
MW: 209.6 g/mol
Boiling Pt: 90…94 °C/13mmHg
Density: 1.33 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00190123
CAS Number: 15996-78-8
UN: 2735
ADR: 8 III