2-(Trifluoromethyl)pyridine-5-carboxaldehyde 97%
Supplier: Apollo Scientific
Danger
Synonyms: 6-(Trifluoromethyl)nicotinaldehyde
PC4333-1G PC4333-25G PC4333-5G
APOSPC4333-1GEA 21.9 EUR
APOSPC4333-1G APOSPC4333-25G APOSPC4333-5G
2-(Trifluoromethyl)pyridine-5-carboxaldehyde 97%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde
Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Boiling Pt: 72…74 °C/0.45mmHg
Melting Pt: 52…60 °C
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862647
CAS Number: 386704-12-7
UN: 2811
ADR: 6.1,III