2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,2-Trifluoro-1-(3-(trifluoromethyl)phenyl)ethanone
PC4349-25G PC4349-1G PC4349-5G PC4349-100G PC4349-500G
APOSPC4349-25GEA 288 EUR
APOSPC4349-25G APOSPC4349-1G APOSPC4349-5G APOSPC4349-100G APOSPC4349-500G
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone 97%
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₄F₆O
Boiling Pt: 65…67 °C/24mmHg
Density: 1.418 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00061275
CAS Number: 721-37-9
UN: 1224
ADR: 3,III