3-Fluoro-4-nitroaniline 98%
Supplier: Apollo Scientific
Warning
PC4371-25G PC4371-5G
APOSPC4371-25GEA 72.5 EUR
APOSPC4371-25G APOSPC4371-5G
3-Fluoro-4-nitroaniline 98%
3-Fluoro-4-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412239
CAS Number: 2369-13-3
EINECS: 219-130-6