2'-Fluoro-4'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Fluoro-4-(trifluoromethyl)phenyl)ethanone
PC4371L-1G PC4371L-25G PC4371L-5G
APOSPC4371L-1GEA 31.9 EUR
APOSPC4371L-1G APOSPC4371L-25G APOSPC4371L-5G
2'-Fluoro-4'-(trifluoromethyl)acetophenone
2'-Fluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD00061247
CAS Number: 122023-29-4