3'-Fluoro-5'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[3-Fluoro-5-(trifluoromethyl)phenyl]ethan-1-one, 3-Acetyl-5-fluorobenzotrifluoride
PC4371O-25G PC4371O-50G PC4371O-5G
APOSPC4371O-25GEA 173 EUR
APOSPC4371O-25G APOSPC4371O-50G APOSPC4371O-5G
3'-Fluoro-5'-(trifluoromethyl)acetophenone 98%
3'-Fluoro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MW: 206.1370 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061250
CAS Number: 202664-54-8