2-Fluoro-6-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC4372N-25G PC4372N-100G PC4372N-500G
APOSPC4372N-25GEA 58 EUR
APOSPC4372N-25G APOSPC4372N-100G APOSPC4372N-500G
2-Fluoro-6-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 156 °C
Density: 1.432 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00061273
CAS Number: 60611-24-7