3-Fluoro-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-fluoro-5-(trifluoromethyl)benzaldehyde
PC4372O-100G PC4372O-25G PC4372O-5G
APOSPC4372O-100GEA 324 EUR
APOSPC4372O-100G APOSPC4372O-25G APOSPC4372O-5G
3-Fluoro-5-(trifluoromethyl)benzaldehyde 97%
3-Fluoro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 175 °C
Density: 1.376 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00061312
CAS Number: 188815-30-7