4-Fluoro-2-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC4372P-25G PC4372P-5G PC4372P-100G
APOSPC4372P-25GEA 50.9 EUR
APOSPC4372P-25G APOSPC4372P-5G APOSPC4372P-100G
4-Fluoro-2-(trifluoromethyl)benzaldehyde 97%
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 113…115 °C
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00061313
CAS Number: 90176-80-0
UN: 1989
ADR: 3 III