4-Fluoro-3-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC4372Q-100G PC4372Q-25G PC4372Q-5G
APOSPC4372Q-100GEA 134 EUR
APOSPC4372Q-100G APOSPC4372Q-25G APOSPC4372Q-5G
4-Fluoro-3-(trifluoromethyl)benzaldehyde 97%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 60 °C/0.75mmHg
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00061314
CAS Number: 67515-60-0