α,α,α,4-Tetrafluoro-o-toluic acid 98%
Supplier: Apollo Scientific
Warning
PC4373D-100G PC4373D-25G PC4373D-5G
APOSPC4373D-100GEA 126 EUR
APOSPC4373D-100G APOSPC4373D-25G APOSPC4373D-5G
α,α,α,4-Tetrafluoro-o-toluic acid 98%
α,α,α,4-Tetrafluoro-o-toluic acid
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Boiling Pt: 251
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00040982
CAS Number: 141179-72-8