2-Fluoro-6-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Cyano-3-fluorobenzotrifluoride
PC4373H-1G PC4373H-25G PC4373H-5G
APOSPC4373H-1GEA 21.9 EUR
APOSPC4373H-1G APOSPC4373H-25G APOSPC4373H-5G
2-Fluoro-6-(trifluoromethyl)benzonitrile 97%
2-Fluoro-6-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Melting Pt: 26…28 °C
Density: 1.373 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00061153
CAS Number: 133116-83-3
UN: 3439
ADR: 6.1,III