2-Fluoro-3-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-3-(trifluoromethyl)benzene-1-carbonyl chloride
PC4374M-25G PC4374M-5G
APOSPC4374M-25GEA 149 EUR
APOSPC4374M-25G APOSPC4374M-5G
2-Fluoro-3-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-3-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
Boiling Pt: 192…194 °C
Density: 1.517 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00061154
CAS Number: 208173-19-7
UN: 3265
ADR: 8 III