2-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC4374O-100G PC4374O-25G PC4374O-5G
APOSPC4374O-100GEA 358 EUR
APOSPC4374O-100G APOSPC4374O-25G APOSPC4374O-5G
2-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-5-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
Boiling Pt: 190…191 °C
Density: 1.499 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00061156
CAS Number: 207981-46-2
UN: 3265
ADR: 8,II