3-Fluoro-5-(trifluoromethyl)benzylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: (3-Fluoro-5-(trifluoromethyl)phenyl)methanamine
PC4374X-1G PC4374X-5G
APOSPC4374X-1GEA 29 EUR
APOSPC4374X-1G APOSPC4374X-5G
3-Fluoro-5-(trifluoromethyl)benzylamine 97%
3-Fluoro-5-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
MW: 193.14 g/mol
Density: 1.319 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00061163
CAS Number: 150517-77-4
UN: 2735
ADR: 8,II